Skip to Content
Merck
All Photos(1)

Key Documents

654035

Sigma-Aldrich

Poly(dichlorophosphazene)

a polyphosphazene precursor

Synonym(s):

PDCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cl2NP)x
CAS Number:
MDL number:
UNSPSC Code:
12352103

refractive index

n20/D 1.578 (lit.)

density

1.823 g/mL at 25 °C (lit.)

storage temp.

−20°C

General description

Poly [(organo) phosphazene] polymers have an inorganic backbone made up of alternating nitrogen and phosphorous atoms. The inorganic atoms are connected by alternating single and double bonds.

Application

Poly(dichlorophosphazene) (PDCP) may be used in the synthesis of higher biologically active organophosphazene polymers.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

108.9 °F

Flash Point(C)

42.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (dichlorophosphazene) as a precursor for biologically active polyphosphazenes: synthesis, characterization, and stabilization.
Andrianov A K, et al.
Marcromolecules, 37(2), 414-420 (2004)
Ambient temperature synthesis of poly (dichlorophosphazene) with molecular weight control.
Honeyman C H, et al.
Journal of the American Chemical Society, 117(26), 7035-7036 (1995)
Applicative aspects of poly(organophosphazenes) (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service