Skip to Content
Merck
All Photos(1)

Key Documents

654019

Sigma-Aldrich

N-Boc-cis-4-hydroxy-L-proline

97%

Synonym(s):

N-Boc-cis-4-hydroxypyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO5
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D -50.0±3°, c = 0.67 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

146 °C (D) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1C[C@@H](O)C[C@H]1C(O)=O

InChI

1S/C10H17NO5/c1-10(2,3)16-9(15)11-5-6(12)4-7(11)8(13)14/h6-7,12H,4-5H2,1-3H3,(H,13,14)/t6-,7-/m0/s1

InChI key

BENKAPCDIOILGV-BQBZGAKWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service