Skip to Content
Merck
All Photos(1)

Documents

652652

Sigma-Aldrich

2-Bromobenzaldehyde ethylene acetal

96%

Synonym(s):

2-(2-Bromophenyl)-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BrO2
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.569 (lit.)

density

1.535 g/mL at 25 °C (lit.)

SMILES string

Brc1ccccc1C2OCCO2

InChI

1S/C9H9BrO2/c10-8-4-2-1-3-7(8)9-11-5-6-12-9/h1-4,9H,5-6H2

InChI key

IWSGKSUCFVOWQU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>249.8 °F

Flash Point(C)

> 121 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polina Novoseltseva et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(10), 2276-2284 (2020-01-04)
New chiral N,C-chelate organoboron compounds based on benzylideneamines (bza) with the general formula of B(bza-R)Mes2 (R=H or Me; Mes=mesityl) are reported. A chiral substituent group R- or S-CH(CH3 )Ph (Ph=phenyl) was introduced to the imine center, which imposed a previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service