Skip to Content
Merck
All Photos(1)

Documents

652415

Sigma-Aldrich

Iodomethane-13C,d

≥98 atom % D, ≥99 atom % 13C, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methyl-13C,d iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D13CH2I
CAS Number:
Molecular Weight:
143.94
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

≥99 atom % 13C
≥98 atom % D

Assay

≥99% (CP)

contains

copper as stabilizer

bp

42 °C (lit.)

mp

-66.5 °C

density

2.312 g/mL at 25 °C

application(s)

environmental

mass shift

M+2

storage temp.

2-8°C

SMILES string

[2H][13CH2]I

InChI

1S/CH3I/c1-2/h1H3/i1+1D

InChI key

INQOMBQAUSQDDS-VVKOMZTBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service