Skip to Content
Merck
All Photos(1)

Key Documents

647780

Sigma-Aldrich

Silicon

wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Silicon element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

crystalline (cubic (a = 5.4037))
wafer (single side polished)

contains

phosphorus as dopant

resistivity

1-10 Ω-cm

diam. × thickness

2 in. × 0.5 mm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

semiconductor properties

<100>, N-type

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

0 vortex defects. Etch pitch density (EPD) < 100 (cm-2). Resistivity 10-3 - 40 Ωcm
Oxygen content: <= 1~1.8 x 1018 /cm3; Carbon content: <= 5 x 1016 /cm3; Boule diameter: 1~8 ″

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pil Ju Ko et al.
Journal of nanoscience and nanotechnology, 13(4), 2451-2460 (2013-06-15)
The physical properties of porous materials are being exploited for a wide range of applications including optical biosensors, waveguides, gas sensors, micro capacitors, and solar cells. Here, we review the fast, easy and inexpensive electrochemical anodization based fabrication porous silicon
Hyunhui Kim et al.
Journal of nanoscience and nanotechnology, 13(5), 3559-3563 (2013-07-19)
Silicon sheets were fabricated by a new fabricating method, spin casting with various rotation speeds of the graphite mold. The microstructure of spin-cast silicon sheets were investigated using an electron probe microanalyzer (EPMA) and scanning electron microscope/electron backscatter diffraction/orientation image
Jaewoo Lee et al.
Journal of nanoscience and nanotechnology, 13(5), 3495-3499 (2013-07-19)
A spin-casting process for fabricating polycrystalline silicon sheets for use as solar cell wafers is proposed, and the parameters that control the sheet thickness are investigated. A numerical study of the fluidity of molten silicon indicates that the formation of
Jae Cheol Shin et al.
Journal of nanoscience and nanotechnology, 13(5), 3511-3514 (2013-07-19)
We have characterized the structural properties of the ternary In(x)Ga(1-x)As nanowires (NWs) grown on silicon (Si) substrates using metalorganic chemical vapor deposition (MOCVD). Au catalyzed vapor-liquid-solid (VLS) mode was used for the NW growth. The density of the In(x)Ga(1-x)As NW
Youngin Jeon et al.
Journal of nanoscience and nanotechnology, 13(5), 3350-3353 (2013-07-19)
Si-nanowire (NW)-array-based NOT-logic circuits were constructed on plastic substrates. The Si-NW arrays were fabricated on a Si wafer through top-down methods, including conventional photolithography and crystallographic wet etching, and transferred onto the plastic substrates. Two field-effect transistors were fabricated on

Articles

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service