Skip to Content
Merck
All Photos(1)

Documents

646423

Sigma-Aldrich

3-Hydroxypicolinamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

193-197 °C (lit.)

SMILES string

NC(=O)c1ncccc1O

InChI

1S/C6H6N2O2/c7-6(10)5-4(9)2-1-3-8-5/h1-3,9H,(H2,7,10)

InChI key

VIHUZJYFQOEUMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Clays et al.
Biochemistry, 30(29), 7271-7276 (1991-07-23)
Using multifrequency phase fluorometry, fluorescence lifetimes have been assigned to the different protolytic forms of the antibiotic virginiamycin S. These lifetimes are 0.476 +/- 0.005 ns for the uncharged form, 1.28 +/- 0.2 and 7.4 +/- 0.2 ns for the
Ieda Lucia Viana Rosa et al.
Journal of fluorescence, 21(4), 1575-1583 (2011-01-29)
This work reports on the synthesis and characterization of a new complex of Eu(3+) with the 3-hydroxypicolinamide ligand (Hhpa). Here we present an approach for obtaining bis[2-carbamoyl(κO)pyridin-3-olato(κO')] lanthanide complexes, which were characterized through elemental analysis, thermal analysis, infrared and photoluminescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service