Skip to Content
Merck
All Photos(2)

Key Documents

645141

Sigma-Aldrich

2,2′:5′,2″-Terthiophene-5-boronic acid pinacol ester

96%

Synonym(s):

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2′:5′,2″-terthiophene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$617.00

$617.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$617.00

About This Item

Empirical Formula (Hill Notation):
C18H19BO2S3
CAS Number:
Molecular Weight:
374.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$617.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

96%

form

solid

mp

109-113 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(s2)-c3ccc(s3)-c4cccs4

InChI

1S/C18H19BO2S3/c1-17(2)18(3,4)21-19(20-17)16-10-9-15(24-16)14-8-7-13(23-14)12-6-5-11-22-12/h5-11H,1-4H3

InChI key

FIUQKESHHZFUGG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service