Skip to Content
Merck
All Photos(1)

Key Documents

643637

Sigma-Aldrich

Silica, mesostructured

MSU-H (large pore 2D hexagonal)

Synonym(s):

Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

surface area

~750 m2/g , BET

unit cell size

~11.6 nm

pore size

0.91 cm3/g pore volume
~7.1 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

bulk density

0.12 g/mL

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adsorbant, catalyst support, nanoscale template

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
K E Kovalenko et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1048-1060 (2013-08-24)
Boreal wetlands play an important role in global carbon balance. However, their ecosystem function is threatened by direct anthropogenic disturbance and climate change. Oil sands surface mining in the boreal regions of Western Canada denudes tracts of land of organic
C J Horwell et al.
Environmental research, 127, 63-73 (2013-11-26)
The six week eruption of Eyjafjallajökull volcano in 2010 produced heavy ash fall in a sparsely populated area of southern and south eastern Iceland and disrupted European commercial flights for at least 6 days. We adopted a protocol for the
Yang Ding et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10123-10128 (2013-06-05)
Undulatory locomotion, a gait in which thrust is produced in the opposite direction of a traveling wave of body bending, is a common mode of propulsion used by animals in fluids, on land, and even within sand. As such, it
Yaswanth Kuthati et al.
Journal of nanoscience and nanotechnology, 13(4), 2399-2430 (2013-06-15)
The advent of Nanotechnology has paved a way for improved disease treatment strategies, the most noteworthy being the mesoporous silica nanoparticles (MSNs) which have gained much recent attention in the field of cancer therapy and its diagnosis. The flaws of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service