643629
Aluminosilicate, mesostructured
Al-MSU-F (cellular foam)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(SiO2)x(Al2O3)y
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form
powder
composition
aluminum, 3.0% (mol)
pore size
2.03 cm3/g pore volume
application(s)
battery manufacturing
SMILES string
O=[Si]=O.O=[Al]O[Al]=O
InChI
1S/2Al.O2Si.3O/c;;1-3-2;;;
InChI key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Adsorbent, acid catalyst, catalyst support, nanoscale template
Formula variant
(SiO2)0.9875(Al2O3)0.0125·xH2O
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mery Montinaro et al.
Life sciences, 92(17-19), 903-910 (2013-04-09)
Oxidative stress is considered one of the main events that lead to aging and neurodegeneration. Antioxidant treatments used to counteract oxidative damage have been associated with a wide variety of side effects or at the utmost to be ineffective. The
Danny Verboekend et al.
Chimia, 67(5), 327-332 (2013-07-19)
This review emphasizes key recent accomplishments towards the industrial exploitation of hierarchically structured zeolites in catalytic processes. A major milestone comprises the demonstration that affordable post-synthetic modifications enable the transformation of any conventional zeolite into hierarchical analogues with tunable porosity
Ke Zhang et al.
Chemical communications (Cambridge, England), 49(31), 3245-3247 (2013-03-15)
Alcohol (methanol, ethanol, 1-propanol, 2-propanol and 1-butanol) and water vapor adsorption in zeolitic imidazolate frameworks (ZIF-8, ZIF-71 and ZIF-90) with similar crystal sizes was systematically studied. The feasibility of applying these ZIF materials to the recovery of bio-alcohols is evaluated
Gas separation: Trapdoors in zeolites.
Trees De Baerdemaeker et al.
Nature chemistry, 5(2), 89-90 (2013-01-25)
Liling Zhang et al.
Journal of the American Chemical Society, 135(9), 3722-3728 (2013-02-22)
A new force field and a hybrid Monte Carlo/molecular dynamics simulation method are developed to investigate the structural transition of zeolitic imidazolate framework-8 (ZIF-8) induced by N2 sorption. At a high loading (approximately 50 N2 molecules per unit cell), ZIF-8
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service