Skip to Content
Merck
All Photos(1)

Documents

643610

Sigma-Aldrich

Bis(1-benzotriazolyl)methanethione

97%

Synonym(s):

1,1′-(Thiocarbonyl)bis-1H-benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N6S
CAS Number:
Molecular Weight:
280.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

170-174 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C(n1nnc2ccccc12)n3nnc4ccccc34

InChI

1S/C13H8N6S/c20-13(18-11-7-3-1-5-9(11)14-16-18)19-12-8-4-2-6-10(12)15-17-19/h1-8H

InChI key

ZRXHYHZENMJKMG-UHFFFAOYSA-N

Application

Reactant used as a thioacylating agent

Reactant involved in:


  • Multicomponent heterocyclization reactions
  • Synthesis of substituted thiosemicarbazides and N-hydroxythioureas
  • Guanylation of amines

Other Notes

Thiophosgene equivalent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service