Skip to Content
Merck
All Photos(1)

Key Documents

643548

Sigma-Aldrich

2-Bromo-3-(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BrF3N
CAS Number:
Molecular Weight:
225.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

37-41 °C

SMILES string

FC(F)(F)c1cccnc1Br

InChI

1S/C6H3BrF3N/c7-5-4(6(8,9)10)2-1-3-11-5/h1-3H

InChI key

OFGSIPQYQUVVPL-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service