Skip to Content
Merck
All Photos(1)

Documents

643394

Sigma-Aldrich

Titanium(III) chloride solution

~10 wt. % in 20-30 wt. % hydrochloric acid

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solution

reaction suitability

reagent type: catalyst
core: titanium

concentration

~10 wt. % in 20-30 wt. % hydrochloric acid

density

1.192 g/mL at 25 °C

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

For use in CDC approved procedures.

Other Notes

Exact concentration stated on label

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-ichi Matsuo et al.
Organic letters, 12(14), 3266-3268 (2010-06-24)
N-p-Toluenesulfonyl (Ts) aldimines reacted with 3-ethoxycyclobutanones by catalysis of titanium(IV) chloride to afford 2,3-di- or 2,3,3-trisubstituted N-Ts-2,3-dihydro-4-pyridones. Synthesis of (+/-)-bremazocine was efficiently accomplished by using this method.
Palaniappan Kulanthaivel et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(9), 966-972 (2004-08-21)
Phase I oxidative metabolism of nitrogen-containing drug molecules to their corresponding N-oxides is a common occurrence. There are instances where liquid chromatography/tandem mass spectometry techniques are inadequate to distinguish this pathway from other oxidation processes, including C-hydroxylations and other heteroatom
L Messori et al.
FEBS letters, 442(2-3), 157-161 (1999-02-03)
The reaction of human serum apotransferrin with titanium(IV) citrate under physiological conditions results in the formation of a specific bis-titanium(IV) transferrin adduct (Ti2Tf hereafter) with two titanium(IV) ions loaded at the iron binding sites. The same specific Ti2Tf complex is
H A Akers et al.
Analytical biochemistry, 154(2), 559-563 (1986-05-01)
Thirteen model alpha-ketocarboxylic acids and alpha-dicarbonyl compounds have been observed to be reduced by titanium(III) chloride. The products of these reactions were shown by melting (boiling) points, mixed melting points, derivatives, refractive indices, infrared, and NMR comparisons with authentic compounds
Gema Ruano et al.
The Journal of organic chemistry, 68(5), 2024-2027 (2003-03-01)
Enantiomerically pure N-substituted epoxyalkene-2-azetidinones reacted with titanocene monochloride to give stereospecifically polyfunctionalized bicyclic beta-lactams. Four isomeric epoxyaldehydes 2 reacted with TiCp2Cl to give exclusively the respective carbacephams 7 while under the same reaction conditions the epoxyesters 1, which are more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service