Skip to Content
Merck
All Photos(1)

Key Documents

642444

Sigma-Aldrich

2-Amino-4-(3,4-difluorophenyl)thiazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$106.00

About This Item

Empirical Formula (Hill Notation):
C9H6F2N2S
CAS Number:
Molecular Weight:
212.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

121-125 °C (lit.)

functional group

fluoro

SMILES string

Nc1nc(cs1)-c2ccc(F)c(F)c2

InChI

1S/C9H6F2N2S/c10-6-2-1-5(3-7(6)11)8-4-14-9(12)13-8/h1-4H,(H2,12,13)

InChI key

NDCSJUJQMRFHEX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahul Hameed P et al.
Scientific reports, 8(1), 7263-7263 (2018-05-10)
The mechanism of efflux is a tour-de-force in the bacterial armoury that has thwarted the development of novel antibiotics. We report the discovery of a novel chemical series with potent antibacterial properties that was engineered to overcome efflux liability. Compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service