Skip to Content
Merck
All Photos(1)

Key Documents

64205

Sigma-Aldrich

3-(Trichlorosilyl)propyl methacrylate

contains BHT as stabilizer, technical, ≥90% (GC)

Synonym(s):

(3-Methacryloyloxypropyl)trichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11Cl3O2Si
CAS Number:
Molecular Weight:
261.61
Beilstein:
1942074
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical

Assay

≥90% (GC)

contains

BHT as stabilizer

refractive index

n20/D 1.465 (lit.)

density

1.239 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCC[Si](Cl)(Cl)Cl

InChI

1S/C7H11Cl3O2Si/c1-6(2)7(11)12-4-3-5-13(8,9)10/h1,3-5H2,2H3

InChI key

DOGMJCPBZJUYGB-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Throm Quinlan et al.
PloS one, 6(8), e23272-e23272 (2011-08-23)
Cells have the ability to actively sense their mechanical environment and respond to both substrate stiffness and stretch by altering their adhesion, proliferation, locomotion, morphology, and synthetic profile. In order to elucidate the interrelated effects of different mechanical stimuli on
Eric J Leaman et al.
ACS synthetic biology, 7(4), 1030-1042 (2018-03-27)
Bacteria utilize diffusible signals to regulate population density-dependent coordinated gene expression in a process called quorum sensing (QS). While the intracellular regulatory mechanisms of QS are well-understood, the effect of spatiotemporal changes in the population configuration on the sensitivity and
Kou Tsukagoshi et al.
Dental materials journal, 39(5), 844-854 (2020-05-26)
The purposes of this study were to evaluate the shear bond strength and perform the computational analysis for silane coupling treatments on the adhesion between CAD/CAM composite resin and resin cement. As silane coupling agents, γ-methacryloyloxy propyltrimethoxysilane, 8-methacryloyloxyoctyl trimethoxysilane, and
Anastasia M Moysenovich et al.
Neuroreport, 31(10), 770-775 (2020-05-30)
Neural transplantation is a promising modality for treatment of neurodegenerative diseases, traumatic brain injury and stroke. Biocompatible scaffolds with optimized properties improve the survival of transplanted neural cells and differentiation of progenitor cells into the desired types of neurons. Silk
Juhong Ahn et al.
Carbohydrate polymers, 217, 160-167 (2019-05-14)
Polysaccharides can be modified by reactive functional groups to enable chemical crosslinking. We studied how different methods of crosslinking methacrylate-functionalized chitosan affected the network structures and various properties relevant for utilization of the chemically crosslinked hydrogels in biomedical applications, including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service