Skip to Content
Merck
All Photos(1)

Key Documents

641677

Sigma-Aldrich

Holmium(III) phosphate

Synonym(s):

Phosphoric acid holmium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HoPO4
CAS Number:
Molecular Weight:
259.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

reaction suitability

reagent type: catalyst
core: holmium

SMILES string

[Ho+3].[O-]P([O-])([O-])=O

InChI

1S/Ho.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3

InChI key

ZFEZZRFBMNCXLQ-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wouter Bult et al.
Pharmaceutical research, 29(3), 827-836 (2011-11-10)
The clinical application of holmium acetylacetonate microspheres (HoAcAcMS) for the intratumoral radionuclide treatment of solid malignancies requires a thorough understanding of their stability. Therefore, an in vitro and an in vivo stability study with HoAcAcMS was conducted. HoAcAcMS, before and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service