Skip to Content
Merck
All Photos(2)

Key Documents

641588

Sigma-Aldrich

Dodecyl methyl sulfoxide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H28OS
CAS Number:
Molecular Weight:
232.43
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

59-63 °C (lit.)

functional group

sulfoxide

SMILES string

CCCCCCCCCCCCS(C)=O

InChI

1S/C13H28OS/c1-3-4-5-6-7-8-9-10-11-12-13-15(2)14/h3-13H2,1-2H3

InChI key

CJPDBKNETSCHCH-UHFFFAOYSA-N

General description

Dodecyl methyl sulfoxide can be synthesized via oxidation of dodecyl methyl sulfide with sodium periodate.[1]

Application

Dodecyl methyl sulfoxide can be used as a substitute for dimethyl sulfoxide in Swern oxidation[2] of primary allylic and benzylic alcohols to the corresponding aldehydes and secondary alcohols to the corresponding ketones.[1][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"New odourless protocols for efficient Pauson?Khand annulations"
Brown.AJ, et al.
Organic & Biomolecular Chemistry, 3(13), 2396-2398 (2005)
"New odorless method for the Corey?Kim and Swern oxidations utilizing dodecyl methyl sulfide (Dod-S-Me)"
Ohsugi I-S, et al.
Tetrahedron, 59(42), 8393-8398 (2003)
"New non-volatile and odorless organosulfur compounds anchored on ionic liquids. Recyclable reagents for Swern oxidation"
He X and Chan HT
Tetrahedron, 62(14), 3389-3394 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service