All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9ClO2
CAS Number:
Molecular Weight:
196.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
solid
SMILES string
COc1cc2C(=O)CCc2cc1Cl
InChI
1S/C10H9ClO2/c1-13-10-5-7-6(4-8(10)11)2-3-9(7)12/h4-5H,2-3H2,1H3
InChI key
XIUZEIYKEBEXIB-UHFFFAOYSA-N
Application
Reactant for:
- Preparation of azaheterocycles via a palladium(II)-catalyzed ring-expansion reaction
- Preparation of phenyldihydroindene hydroxy derivatives as dopamine receptor ligands
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service