Skip to Content
Merck
All Photos(3)

Key Documents

637939

Sigma-Aldrich

1-Bromo-2,4,6-triisopropylbenzene

95%

Synonym(s):

2,4,6-Triisopropylbromobenzene, 2-Bromo-1,3,5-triisopropylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$47.10
25 G
$224.00
100 G
$536.00

$47.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$47.10
25 G
$224.00
100 G
$536.00

About This Item

Empirical Formula (Hill Notation):
C15H23Br
CAS Number:
Molecular Weight:
283.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$47.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

95%

refractive index

n20/D 1.523 (lit.)

bp

96-97 °C/0.1 mmHg (lit.)

density

1.118 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CC(C)c1cc(C(C)C)c(Br)c(c1)C(C)C

InChI

1S/C15H23Br/c1-9(2)12-7-13(10(3)4)15(16)14(8-12)11(5)6/h7-11H,1-6H3

InChI key

FUMMYHVKFAHQST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Bromo-2,4,6-triisopropylbenzene can be used in the synthesis of 1-fluoro-2,4,6-triisopropylbenzene, a fluorine-substituted aromatic building block for agrochemical synthesis and pharmaceutical production.[1] It can also be used in the synthesis of di-2,4,6-triisopropylphenyl diselenide via reaction with metallic selenium in the presence of tertiary-butyl lithium.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Stereoselective synthesis of (E)-1-iodo-1-selenoalkenes via hydroalumination?iodination of 1-alkynyl selenides"
Balado-Perez S, et al
Tetrahedron Letters, 46(29), 4883-4886 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service