Skip to Content
Merck
All Photos(2)

Key Documents

637475

Sigma-Aldrich

6-Amino-3-pyridinecarbonitrile

97%

Synonym(s):

2-Amino-5-cyanopyridine, 6-Aminonicotinonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$57.00

$57.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$57.00

About This Item

Linear Formula:
C5H3NNH2CN
CAS Number:
Molecular Weight:
119.12
Beilstein:
113876
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

159-163 °C (lit.)

functional group

nitrile

SMILES string

Nc1ccc(cn1)C#N

InChI

1S/C6H5N3/c7-3-5-1-2-6(8)9-4-5/h1-2,4H,(H2,8,9)

InChI key

KDVBYUUGYXUXNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service