Skip to Content
Merck
All Photos(1)

Key Documents

635650

Sigma-Aldrich

2-Iodobenzylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4ICH2ZnBr
CAS Number:
Molecular Weight:
362.32
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

impurities

1-3% inorganic chloride and iodide

density

1.025 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]Cc1ccccc1I

InChI

1S/C7H6I.BrH.Zn/c1-6-4-2-3-5-7(6)8;;/h2-5H,1H2;1H;/q;;+1/p-1

InChI key

CYLNVPVVESOXDT-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service