Skip to Content
Merck
All Photos(1)

Key Documents

633429

Sigma-Aldrich

3,5-Dimethylpyrazole-1-carboxamidine hydrochloride, polymer-supported

100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

1.0-2.0 mmol/g loading

particle size

100-200 mesh

InChI

1S/C6H10N4.Cl/c1-4-3-5(2)10(9-4)6(7)8;/h3H,1-2H3,(H3,7,8);

InChI key

VEBVDVIHRVPBQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymer-bound reagent for amidation of amines to make guanidines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Pogány et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(4), 1466-1473 (2008-06-20)
In the paper a joint experimental and theoretical study of 3,5-dimethyl-1H-pyrazole-1-carboxamidine (L) as well as its complexes CoL2(H2O)2(NO3)2 and NiL2(H2O)2(NO3)2 is reported. On the basis of FT-IR experiments and a DFT-derived scaled quantum mechanical force field the normal coordinate analysis
L Spero et al.
Biochimica et biophysica acta, 251(3), 345-356 (1971-12-28)
Guanidination of the free amino groups of staphylococcal enterotoxin B with 3,5-dimethyl-1-guanylpyrazole converted 31-32 of 33 epsilon-amino groups and 30% of the N-terminal residue. This product, although markedly reduced in solubility, suffered no gross change in conformation and retained full
Antonia G Miller et al.
Annals of the New York Academy of Sciences, 1043, 195-200 (2005-07-23)
Protein cross-linking via the Maillard reaction with alpha-dicarbonyl compounds has been the subject of intense scrutiny in the literature. We report here a study of the impact of this cross-linking on enzyme function. Protein function following glycation was examined by
A Bernhardt et al.
The journal of peptide research : official journal of the American Peptide Society, 50(2), 143-152 (1997-08-01)
Peptide-4-nitroanilides can be quickly synthesised using an Fmoc-based approach on 2-chlorotritylchloride resin. Preformed building blocks Fmoc-Xaa-NH-Np (Xaa = Cit, Cys, Gln, His, Lys, Orn, Ser, Thr, Tyr, Trp) can be attached via side chain to the 2-chlorotritylchloride linker of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service