Skip to Content
Merck
All Photos(4)

Key Documents

632309

Sigma-Aldrich

Magnesium hydroxide

nanopowder, <100 nm particle size (laser PSA), 99.8% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8% trace metals basis

form

nanopowder

surface area

94.8 m2/g

particle size

<100 nm (laser PSA)
5.8-20 nm (TEM)
<50 nm (XRD)

mp

350 °C (lit.)

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tengfei Lv et al.
Journal of hazardous materials, 237-238, 121-132 (2012-09-11)
The present study explores the potential of Mg-Ca-Al (NO(3)) hydrotalcite-like compounds (MgCaAlNO(3)-HTlcs) for the removal of fluoride from protein solutions. In this study, the Mg(3-x)Ca(x)AlNO(3)-HTlcs (x=0-3, x is the mol.% of Ca) were synthesized and characterized by SEM, XRD, FTIR
Anna L Harrison et al.
Environmental science & technology, 47(1), 126-134 (2012-07-10)
Atmospheric CO(2) is sequestered within ultramafic mine tailings via carbonation of Mg-bearing minerals. The rate of carbon sequestration at some mine sites appears to be limited by the rate of CO(2) supply. If carbonation of bulk tailings were accelerated, large
Liang Wang et al.
Cancer, 119(2), 348-355 (2012-07-20)
Extranodal natural killer/T-cell lymphoma, nasal type (ENKTL) is a distinct subtype of non-Hodgkin lymphoma in which the upper aerodigestive tract is the most commonly involved site. To date, optimal treatment strategies and prognosis for patients with ENKTL have not been
Guang Liu et al.
Nanoscale, 5(3), 1074-1081 (2012-12-21)
Highly crumpled graphene nanosheets (GNS) with a BET surface area as high as 1159 m(2) g(-1) was fabricated by a thermal exfoliation method. A systematic investigation was performed on the hydrogen sorption properties of MgH(2)-5 wt% GNS nanocomposites acquired by
Tomohito Kameda et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(1), 86-91 (2012-10-04)
A selective catalytic reduction process is generally used to treat NO(x); however, this approach results in the leakage of ammonia and requires expensive catalysts. In light of these issues, the specific objective of this study was to develop a new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service