Skip to Content
Merck
All Photos(1)

Key Documents

631574

Sigma-Aldrich

(S)-(−)-2,2′-Bis(methoxymethoxy)-1,1′-binaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2OC10H6)2
CAS Number:
Molecular Weight:
374.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D -90°, c = 1% in chloroform

mp

100-104 °C (lit.)

InChI

1S/C24H22O4/c1-25-15-27-21-13-11-17-7-3-5-9-19(17)23(21)24-20-10-6-4-8-18(20)12-14-22(24)28-16-26-2/h3-14H,15-16H2,1-2H3

InChI key

YIAQRNNJNMLGTP-UHFFFAOYSA-N

Application

3- or 3,3′-di-substituted products can be obtained easily via ortho-metallation and subsequent treatment with an electrophile to give the corresponding BINOL derivatives.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cox, P. J. et al
Tetrahedron Letters, 33, 2253-2253 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service