Skip to Content
Merck
All Photos(1)

Key Documents

630217

Sigma-Aldrich

Poly(fluorescein isothiocyanate allylamine hydrochloride)

Poly(allylamine hydrochloride) : Fluorescein isothiocyanate 50:1

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$756.00

$756.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$756.00

About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$756.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

solubility

H2O: soluble

λmax

494 nm

Looking for similar products? Visit Product Comparison Guide

General description

Poly(allylamine hydrochloride) : Fluorescein isothiocyanate 50:1

Other Notes

Compound was made using Poly(allylamine hydrochloride), polymer base Mw ca. 15,000

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service