Skip to Content
Merck
All Photos(1)

Key Documents

630012

Sigma-Aldrich

2,2′-Dichlorobenzil

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$138.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$138.00

About This Item

Linear Formula:
ClC6H4COCOC6H4Cl
CAS Number:
Molecular Weight:
279.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

132-136 °C (lit.)

functional group

chloro
ketone

SMILES string

Clc1ccccc1C(=O)C(=O)c2ccccc2Cl

InChI

1S/C14H8Cl2O2/c15-11-7-3-1-5-9(11)13(17)14(18)10-6-2-4-8-12(10)16/h1-8H

InChI key

VOSNNSVWVJFJCR-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service