Skip to Content
Merck
All Photos(1)

Key Documents

62550

Sigma-Aldrich

Lithium iodide hydrate

purum p.a., ≥98.0% (calc. based on dry substance, AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiI · xH2O
CAS Number:
Molecular Weight:
133.85 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥98.0% (calc. based on dry substance, AT)
form:
crystals

grade

purum p.a.

Assay

≥98.0% (calc. based on dry substance, AT)

form

crystals

impurities

≤30% water
2-3 mol water

mp

73 °C (lit.)

density

3.48 g/mL at 25 °C (lit.)

anion traces

bromide, chloride (as Cl-): ≤200 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Li+].[I-].[H]O[H]

InChI

1S/HI.Li.H2O/h1H;;1H2/q;+1;/p-1

InChI key

WAZWGFFJLSIDMX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service