Skip to Content
Merck
All Photos(1)

Key Documents

617458

Sigma-Aldrich

4-Aminobutyric acid-2,2-d2

98 atom % D

Synonym(s):

GABA-d2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$1,600.00

$1,600.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

Select a Size

Change View
250 MG
$1,600.00

About This Item

Linear Formula:
H2N(CH2)2CD2CO2H
CAS Number:
Molecular Weight:
105.13
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,600.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

isotopic purity

98 atom % D

form

solid

availability

not available in Japan

technique(s)

mass spectrometry (MS): suitable

mp

195 °C (dec.) (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(CCN)C(O)=O

InChI

1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)/i2D2

InChI key

BTCSSZJGUNDROE-CBTSVUPCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service