Skip to Content
Merck
All Photos(1)

Key Documents

617210

Sigma-Aldrich

Anthraquinone-d8

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D8O2
CAS Number:
Molecular Weight:
216.26
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

bp

379-381 °C (lit.)

mp

284-286 °C (lit.)

application(s)

electronics

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c2C(=O)c3c([2H])c([2H])c([2H])c([2H])c3C(=O)c2c1[2H]

InChI

1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

RZVHIXYEVGDQDX-PGRXLJNUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service