Skip to Content
Merck
All Photos(1)

Key Documents

616451

Sigma-Aldrich

1,2,4,5-Tetramethylbenzene-d14

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D14
Molecular Weight:
148.30
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

solid

mp

76-80 °C

density

0.924 g/mL

mass shift

M+14

SMILES string

[2H]c1c(c(c([2H])c(c1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C10H14/c1-7-5-9(3)10(4)6-8(7)2/h5-6H,1-4H3/i1D3,2D3,3D3,4D3,5D,6D

InChI key

SQNZJJAZBFDUTD-ZVEBFXSZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenxian Hu et al.
Journal of cellular biochemistry, 120(3), 4113-4120 (2018-10-12)
Long intergenic noncoding RNA-p21 (lincRNA-p21) has been proved in the pathogenesis of aortic aneurysms, while its functionality in thoracic aortic aneurysms (TAA) and the mechanism of function remains unclear. Therefore our study aimed to investigate the role of lincRNA-p21 in
Kazuyuki Nakagome et al.
Journal of immunology (Baltimore, Md. : 1950), 179(1), 284-294 (2007-06-21)
Although eosinophils play an essential role in allergic inflammation, their role has recently been under controversy. Epidemic studies suggest that hypereosinophilia induced by parasite infection could suppress subsequent Ag sensitization, although the mechanism has not been fully clarified. In this
Eduardo Branco de Sousa et al.
Revista brasileira de ortopedia, 57(2), 314-320 (2021-03-31)
Objective  Our goal was to evaluate the modulation of the synovial fluid cells (SFC) from patients with and without osteoarthritis (OA) by bone morphogenetic protein 4 (BMP-4), Smad-3 and transforming growth factor beta (TGF-β). Methods  Synovial fluid was collected from
Yusuke Tamura et al.
Molecular oncology, 15(5), 1289-1307 (2020-12-21)
Development of innovative therapeutic modalities would address an unmet clinical need in the treatment of triple negative breast cancer (TNBC). Activation of retinoic acid-inducible gene-I (RIG-I)-like receptors (RLRs) such as melanoma differentiation-associated gene 5 (MDA5) and RIG-I in cancer cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service