Skip to Content
Merck
All Photos(1)

Key Documents

61550

Sigma-Aldrich

Lanthanum(III) oxide

≥99.98% (metals basis)

Synonym(s):

Dilanthanum trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La2O3
CAS Number:
Molecular Weight:
325.81
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.0% (EDTA 0.1 M, KT)
≥99.98% (metals basis)

form

solid

impurities

≤300 mg/kg total metallic impurities (trace metal analysis (JCPMS))

density

6.51 g/mL at 25 °C (lit.)

cation traces

Ca: ≤50 mg/kg

application(s)

battery manufacturing

SMILES string

O=[La]O[La]=O

InChI

1S/2La.3O

InChI key

KTUFCUMIWABKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to LAMOX fast ion conductors and superconductors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Nogiwa-Valdez et al.
Acta biomaterialia, 9(4), 6226-6235 (2012-12-12)
Zirconia has been used as an orthopaedic material since 1985 and is increasingly used in dental applications. One major concern with the use of zirconia is the significant loss in mechanical properties through hydrothermal degradation, with the uncontrolled transformation of
Pei Lu et al.
Environmental technology, 33(7-9), 1029-1036 (2012-06-23)
In this paper, catalytic samples of 10, 20, 30, 40 and 50% (w/w) urea/activated carbon fibre (AFC), 10% urea--5% La2O3/ACF, 10% urea--10% La2O3/ACF, 10% urea--15% La2O3/ACF, 20% urea--5% La2O3/ACF, 20% urea--10% La2O3/ACF, and 20% urea-15% La2O3/ACF were prepared and used
Timothy J Boyle et al.
Dalton transactions (Cambridge, England : 2003), 39(34), 8050-8063 (2010-07-29)
A novel family of lanthanide zinc carboxylate compounds was synthesized, characterized (structural determination and luminescent behavior), and investigated for utility as single-source precursors to Ln-Zn-O nanoparticles. Carboxylic acids [H-ORc = H-OPc (H-O(2)CCH(CH(3))(2), H-OBc (H-O(2)CC(CH(3))(3), H-ONc (H-O(2)CCH(2)C(CH(3))(3))] were individually reacted with
A K Singh et al.
Optics letters, 37(5), 776-778 (2012-03-02)
Low-power-threshold cw laser-induced incandescence (CWLII) has been observed in La(2)O(3):Er(3+)-Yb(3+) phosphor on excitation by a 976 nm IR laser. It is suggested that incandescence originates from the extensive heating induced by the nonradiative processes taking place following the laser excitation.
Hui Sun et al.
Bioresource technology, 101(3), 953-958 (2009-09-22)
ZrO(2) supported La(2)O(3) catalyst prepared by impregnation method was examined in the transesterification reaction of sunflower oil with methanol to produce biodiesel. It was found that the catalyst with 21 wt% loaded La(2)O(3) and calcined at 600 degrees C showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service