Skip to Content
Merck
All Photos(1)

Key Documents

614289

Sigma-Aldrich

Diethyl-1,1,1′,1′-d4-stilbestrol-3,3′,5,5′-d4 (mixture of E- and Z- forms)

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D8H12O2
Molecular Weight:
276.40
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mp

169-172 °C (lit.)

mass shift

M+8

SMILES string

[2H]c1cc(O)cc([2H])c1\C(=C(\c2c([2H])cc(O)cc2[2H])C([2H])([2H])C)C([2H])([2H])C

InChI

1S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20H,3-4H2,1-2H3/b18-17+/i3D2,4D2,5D,6D,7D,8D

InChI key

RGLYKWWBQGJZGM-AYAXEXGHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service