Skip to Content
Merck
All Photos(1)

Key Documents

614033

Sigma-Aldrich

Lithocholic acid-2,2,3,4,4-d5

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,860.00

$1,860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

Select a Size

Change View
100 MG
$1,860.00

About This Item

Empirical Formula (Hill Notation):
C24D5H35O3
Molecular Weight:
381.60
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

isotopic purity

98 atom % D

form

solid

mp

184-186 °C (lit.)

mass shift

M+5

SMILES string

[2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC[C@@H]2C([2H])([2H])[C@]1([2H])O)[C@H](C)CCC(O)=O

InChI

1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1/i10D2,14D2,17D

InChI key

SMEROWZSTRWXGI-WODFLMNXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bin Zan et al.
Biomedical chromatography : BMC, 34(7), e4835-e4835 (2020-03-22)
Bear bile is a valuable medicinal material used in traditional Chinese medicine for over 2000 years. However, developing a substitute has become necessary because of protection measures for this endangered species. The ingredients of in vitro cultured bear bile powder (CBBP)
Jaime Gea et al.
Journal of separation science, 40(23), 4549-4562 (2017-10-11)
The characterization of ancient lipids from prehistoric sediments (fumiers) located in a rock-selter has been possible after the optimization of an analytical method based on the microwave-assisted extraction and solid-phase extraction clean-up step and a final derivatization step followed by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service