Skip to Content
Merck
All Photos(1)

Key Documents

612030

Supelco

Nicotine solution

NMR reference standard, 2 mg/mL in DMSO-d6 (99.9 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
$3,340.00

$3,340.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
10 EACH
$3,340.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2
Molecular Weight:
162.23
UNSPSC Code:
12142201
NACRES:
NA.24

$3,340.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

NMR reference standard
analytical standard

composition

TMS, 0.05%

concentration

2 mg/mL in DMSO-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

NMR reference standard (2 mg/mL in DMSO-d6 (99.9 atom % D), TMS 0.05%) eliminates the sample preparation uncertainty when evaluating instrument performance and assures proper instrument performance in quantitative applications. These NMR reference standards allow the users to monitor key operational parameters such as pw90, sensitivity, resolution, and line shape.

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service