Skip to Content
Merck

Skip To

608564

Potassium selenocyanate-13C,15N

98 atom % 15N, 99 atom % 13C, 97% (CP)

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
KSe13C15N
Molecular Weight:
146.06
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Isotopic purity:
99 atom % 13C, 98 atom % 15N
Assay:
97% (CP)
Mass shift:
M+2
Form:
solid
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service


isotopic purity

99 atom % 13C, 98 atom % 15N

assay

97% (CP)

form

solid

mp

100 °C (lit.)

mass shift

M+2

SMILES string

[K][Se][13C]#[15N]

InChI

1S/CHNSe.K/c2-1-3;/h3H;/q;+1/p-1/i1+1,2+1;

InChI key

KYEKHFSRAXRJBR-AWQJXPNKSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
775010791024673684
assay

97% (CP)

assay

97% (CP)

assay

98% (CP)

assay

98% (CP)

mass shift

M+2

mass shift

M+2

mass shift

M+2

mass shift

M+2

isotopic purity

99 atom % 13C, 98 atom % 15N

isotopic purity

99 atom % 13C, 98 atom % 15N

isotopic purity

99 atom % 13C, 98 atom % 15N

isotopic purity

99 atom % 13C, 98 atom % 15N

form

solid

form

solid

form

solid

form

solid

mp

100 °C (lit.)

mp

-

mp

-

mp

68 °C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters