Skip to Content
Merck
All Photos(1)

Documents

606014

Sigma-Aldrich

Fumaric acid-13C4

99 atom % 13C, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213C13CH=13CH13CO2H
Molecular Weight:
120.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

299-300 °C (subl.) (lit.)

mass shift

M+4

SMILES string

O[13C](=O)\[13CH]=[13CH]\[13C](O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+/i1+1,2+1,3+1,4+1

InChI key

VZCYOOQTPOCHFL-BHBLSLFXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alex Gomez-Gomez et al.
Journal of pharmaceutical and biomedical analysis, 164, 382-394 (2018-11-23)
Due to its high sensitivity and specificity, liquid chromatography-electrospray tandem mass spectrometry (LC-MS/MS) could be considered as the gold-standard in targeted metabolomics. Although LC-MS/MS allows for the direct detection of a large number of molecules, the proper quantification of highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service