Skip to Content
Merck
All Photos(1)

Key Documents

60402

Sigma-Aldrich

Potassium methoxide solution

~25% in methanol (T)

Synonym(s):

Potassium methylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$225.00
1 L
$610.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$225.00
1 L
$610.00

About This Item

Linear Formula:
CH3OK
CAS Number:
Molecular Weight:
70.13
Beilstein:
3551544
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

concentration

~25% in methanol (T)

refractive index

n20/D 1.37

density

0.95 g/mL at 20 °C

SMILES string

[K+].C[O-]

InChI

1S/CH3O.K/c1-2;/h1H3;/q-1;+1

InChI key

BDAWXSQJJCIFIK-UHFFFAOYSA-N

Application

Potassium methoxide solution in methanol has been used as a base to synthesize imines via aerobic oxidative coupling of benzylic alcohols and amines in the presence of gold nanoparticles supported on titanium dioxide catalyst.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Song et al.
Nature communications, 8, 15322-15322 (2017-05-20)
Nanoparticles are of long-standing interest for the treatment of neurological diseases such as glioblastoma. Most past work focused on methods to introduce nanoparticles into the brain, suggesting that reaching the brain interstitium will be sufficient to ensure therapeutic efficacy. However
Jiying Wen et al.
Biomaterials, 86, 42-55 (2016-02-18)
A library of hyperbranched polyglycerols (HPGs) functionalized with different mole fractions of zwitterionic sulfabetaine and cationic quaternary ammonium ligands was synthesized and characterized. A post-polymerization method was employed that utilized double bond moieties on the dendritic HPG for the coupling
Formation of imines by selective gold-catalysed aerobic oxidative coupling of alcohols and amines under ambient conditions.
Kegnaes S, et al.
Green Chemistry, 12(8), 1437-1441 (2010)
E Ballesteros et al.
Journal of chromatography. A, 719(1), 221-227 (1996-01-05)
An automated gas chromatographic method for the simultaneous determination of cholesterol, alpha-tocopherol and alpha-tocopheryl acetate in edible oils and fats without derivatization is reported. Interferences from lipid material are avoided by using a continuous system to transesterify triglycerides with potassium
Yang Deng et al.
Biomaterials, 35(24), 6595-6602 (2014-05-13)
A key attribute for nanoparticles (NPs) that are used in medicine is the ability to avoid rapid uptake by phagocytic cells in the liver and other tissues. Poly(ethylene glycol) (PEG) coatings has been the gold standard in this regard for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service