Skip to Content
Merck
All Photos(1)

Documents

603775

Sigma-Aldrich

1-Bromo-3-chloropropane-13C3

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl13CH213CH213CH2Br
CAS Number:
Molecular Weight:
160.41
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

bp

144-145 °C (lit.)

density

1.622 g/mL at 25 °C

mass shift

M+3

SMILES string

Cl[13CH2][13CH2][13CH2]Br

InChI

1S/C3H6BrCl/c4-2-1-3-5/h1-3H2/i1+1,2+1,3+1

InChI key

MFESCIUQSIBMSM-VMIGTVKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Repr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>284.0 °F

Flash Point(C)

> 140 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service