Skip to Content
Merck
All Photos(1)

Key Documents

59453

Sigma-Aldrich

(R)-2,2-Dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate

≥99.0% (sum of enantiomers, GC)

Synonym(s):

1,2-Isopropylidene-sn-glycerol 3-tosylate, D-α,β-Isopropylideneglycerol-γ-tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O5S
CAS Number:
Molecular Weight:
286.34
Beilstein:
89800
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]/D −7.0±0.5°, neat

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@H]2COC(C)(C)O2

InChI

1S/C13H18O5S/c1-10-4-6-12(7-5-10)19(14,15)17-9-11-8-16-13(2,3)18-11/h4-7,11H,8-9H2,1-3H3/t11-/m1/s1

InChI key

SRKDUHUULIWXFT-LLVKDONJSA-N

Other Notes

Chiral building block[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.C. Nicolaou et al.
Journal of the Chemical Society. Chemical Communications, 349-349 (1984)
S.K. Bhatia et al.
Tetrahedron Letters, 28, 1729-1729 (1987)
C. Andre et al.
Tetrahedron Asymmetry, 9, 1359-1359 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service