Skip to Content
Merck
All Photos(1)

Key Documents

593478

Sigma-Aldrich

N-(4-Nitrophenyl)formamide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$80.80

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$80.80

About This Item

Empirical Formula (Hill Notation):
C7H6N2O3
CAS Number:
Molecular Weight:
166.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

196-200 °C (lit.)

functional group

amide
nitro

SMILES string

[H]C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H6N2O3/c10-5-8-6-1-3-7(4-2-6)9(11)12/h1-5H,(H,8,10)

InChI key

ZTCQFVRINYOPOH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Birnbaum et al.
Research in experimental medicine. Zeitschrift fur die gesamte experimentelle Medizin einschliesslich experimenteller Chirurgie, 181(2), 105-112 (1982-01-01)
Determination of proteolytic activities gains increasing importance for the diagnosis of various pathologic conditions. Many questions can only be solved with the aid of experiments with animals. In preparation of such studies the pattern of the enzymatic hydrolysis of 20
N E Dixon et al.
Canadian journal of biochemistry, 58(12), 1335-1344 (1980-12-01)
Acetamide and N-methylurea have been shown for the first time to be substrates for jack bean urease. In the enzymatic hydrolysis of urea, formamide, acetamide, and N-methylurea at pH 7.0 and 38 degrees C, kcat has the values 5870, 85

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service