Skip to Content
Merck
All Photos(1)

Key Documents

591505

Sigma-Aldrich

Poly(pentabromophenyl acrylate-co-glycidyl methacrylate)

glycidyl methacrylate 50 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H3Br5O2)m(C7H10O3)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

composition

glycidyl methacrylate, 50 mol %

transition temp

Tg (DSC) 122 °C (onset)

solubility

chloroform: soluble

Looking for similar products? Visit Product Comparison Guide

Application

High refractive index, photosensitive polymer for optical waveguides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service