Skip to Content
Merck
All Photos(1)

Key Documents

58924

Sigma-Aldrich

(−)-Isolongifolene

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(1R)-2,2,7,7-Tetramethyltricyclo[6.2.1.01.6]undec-5-ene

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$146.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$146.00

About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2207559
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D −138±2°, c = 1% in ethanol

refractive index

n20/D 1.499

bp

255-256 °C

density

0.930 g/mL at 20 °C (lit.)

SMILES string

CC1(C)CCC=C2C(C)(C)[C@H]3CC[C@@]12C3

InChI

1S/C15H24/c1-13(2)8-5-6-12-14(3,4)11-7-9-15(12,13)10-11/h6,11H,5,7-10H2,1-4H3/t11-,15-/m0/s1

InChI key

CQUAYTJDLQBXCQ-NHYWBVRUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Isolongifolene is a sesquiterpene that can be found in pine cone and Japanese cedar essential oils.[1]

Application

(-)-Isolongifolene may be used in the preparation of (-)-isolongifolenone, a potent insect repellent.[2]

Other Notes

This (-)-isolongifolene has an enantiomeric purity of 93%; it is thus the highest purity material ever offered commercially[3]; the double bond can be epoxidized[4]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Ranganathan et al.
Tetrahedron, 26, 621-621 (1970)
Isolongifolenone: a novel sesquiterpene repellent of ticks and mosquitoes.
Zhang A, et al.
Journal of Medical Entomology, 46(1), 100-106 (2009)
Suppression of SOS-inducing activity of chemical mutagens by metabolites from microbial transformation of (-)-isolongifolene.
Sakata K, et al.
Journal of Agricultural and Food Chemistry, 58(4), 2164-2167 (2010)
Rienaecker, R. and Graefe, J
Angewandte Chemie (International Edition in English), 97, 348-348 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service