Skip to Content
Merck
All Photos(1)

Key Documents

588822

Sigma-Aldrich

Sodium formate-13C,18O2

95 atom % 18O, 99 atom % 13C

Synonym(s):

Formic acid-13C,18O2 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H13C18O2Na
CAS Number:
Molecular Weight:
73.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C
95 atom % 18O

form

solid

mp

259-262 °C (lit.)

mass shift

M+5

SMILES string

[Na+].[18O-][13CH]=[18O]

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1/i1+1,2+2,3+2;

InChI key

HLBBKKJFGFRGMU-SORTUTTKSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service