Skip to Content
Merck
All Photos(1)

Key Documents

588806

Sigma-Aldrich

Bisphenol A-(dimethyl-d6)

98 atom % D, 99% (CP)

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane-methyl-d6, 4,4-Isopropylidenediphenol-(methyl-D6), 4,4·-Isopropylidenediphenol-(methyl-D6), Deuterated bisphenol A

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,210.00

$1,210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,210.00

About This Item

Empirical Formula (Hill Notation):
C15D6H10O2
CAS Number:
Molecular Weight:
234.32
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

bp

220 °C/4 mmHg (lit.)

mp

157-159 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(c1ccc(O)cc1)(c2ccc(O)cc2)C([2H])([2H])[2H]

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3

InChI key

IISBACLAFKSPIT-WFGJKAKNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruna De Felice et al.
BMC medical genomics, 8, 56-56 (2015-09-09)
Bisphenol A (BPA) is an environmental compounds is known to possess endocrine disruption potentials. Bisphenol A has epigenetic effects as deregulated expression of microRNAs; such epigenetic marks can induce up/down alterations in gene expression that may persist throughout a lifetime.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service