Skip to Content
Merck
All Photos(1)

Key Documents

58825

Sigma-Aldrich

tert-Butyl isocyanoacetate

technical, ≥95% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$203.00
5 ML
$692.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$203.00
5 ML
$692.00

About This Item

Linear Formula:
CNCH2COOC(CH3)3
CAS Number:
Molecular Weight:
141.17
Beilstein:
4130722
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.422

bp

50 °C/0.1 mmHg (lit.)

density

0.97 g/mL at 20 °C (lit.)

functional group

amine
ester
isonitrile

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)C[N+]#[C-]

InChI

1S/C7H11NO2/c1-7(2,3)10-6(9)5-8-4/h5H2,1-3H3

InChI key

IQSYKHVFMYGJER-UHFFFAOYSA-N

Other Notes

Review[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Schollkopf
Angewandte Chemie (International Edition in English), 89, 351-351 (1977)
Stefan Oelmann et al.
Biomacromolecules, 20(1), 90-101 (2018-06-06)
A Passerini three-component polymerization was performed for the synthesis of amphiphilic star-shaped block copolymers with hydrophobic cores and hydrophilic coronae. The degree of polymerization of the hydrophobic core was varied from 5 to 10 repeating units, and the side chain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service