Skip to Content
Merck
All Photos(1)

Key Documents

588083

Sigma-Aldrich

4-Chloro-3-methylphenol-2,6-d2

98 atom % D

Synonym(s):

4-Chloro-m-cresol-2,6-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6D2H(CH3)OH
CAS Number:
Molecular Weight:
144.60
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

bp

235 °C (lit.)

mp

63-65 °C (lit.)

mass shift

M+2

SMILES string

[2H]c1cc(Cl)c(C)c([2H])c1O

InChI

1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3/i2D,4D

InChI key

CFKMVGJGLGKFKI-XRIVEGAOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service