Skip to Content
Merck
All Photos(1)

Key Documents

587133

Sigma-Aldrich

Lutetium(III) bromide

anhydrous, 99.99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LuBr3
CAS Number:
Molecular Weight:
414.68
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99%

form

powder

reaction suitability

reagent type: catalyst
core: lutetium

SMILES string

Br[Lu](Br)Br

InChI

1S/3BrH.Lu/h3*1H;/q;;;+3/p-3

InChI key

DWHGOINJUKABSY-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Santidrián et al.
Physical chemistry chemical physics : PCCP, 22(18), 10063-10075 (2020-04-28)
In the present work, the effect of doping on electronic properties in bulk purified and filled arc-discharge single-walled carbon nanotubes samples is studied for the first time by in situ Raman spectroelectrochemical method. A major challenge to turn the potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service