578932
N-Allyl-N,N-bis(trimethylsilyl)amine
97%
Synonym(s):
(Prop-2-en-1-yl)bis(trimethylsilyl)amine, 1,1,1-Trimethyl-N-2-propen-1-yl-N-(trimethylsilyl)silanamine, 3-Bis(trimethylsilyl)amino-1-propene, Bis(trimethylsilyl)allylamine, N,N-Bis(trimethylsilyl)-2-propen-1-amine, N,N-Bis(trimethylsilyl)allylamine, N-Allyl-1,1,1-trimethyl-N-(trimethylsilyl)silanamine
About This Item
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.440 (lit.)
bp
72 °C/15 mmHg (lit.)
density
0.816 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
C[Si](C)(C)N(CC=C)[Si](C)(C)C
InChI
1S/C9H23NSi2/c1-8-9-10(11(2,3)4)12(5,6)7/h8H,1,9H2,2-7H3
InChI key
CVNCFZIIZGNVFD-UHFFFAOYSA-N
Application
- Complex bicyclic scaffolds by reacting with aryl aldehydes via imine formation followed by efficient multicomponent reactions.
- Dithiasuccinoyl (Dts) heterocycle by reacting with bisthiocarbamoyl chloride.
- Allyl selenide by reacting with aryl selenium salt in the presence of a ruthenium catalyst; Allyl selenide is utilized as a key intermediate in organic synthesis.
AHMDS can also be used as a reagent in hydrosilylation and hydroboration reaction. It may be also used as an electrolyte additive.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
109.0 °F - closed cup
Flash Point(C)
42.78 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service