Skip to Content
Merck
All Photos(2)

Documents

577065

Sigma-Aldrich

Polypyrrole

undoped, extent of labeling: ~20 wt. % loading, composite with carbon black

Synonym(s):

PPy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

extent of labeling

~20 wt. % loading

conductivity

~8.5 S/cm

solubility

H2O: insoluble
organic solvents: insoluble

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polypyrrole (PPy) is a conductive and highly stable polymer. It may be prepared by a standard electrochemical technique. PPy may also be prepared by reacting ß-napthalene sulfonic acid (NSA) and ammonium peroxo-disulphate in aqueous medium. The charges on the surfaces can be easily modified by doping the polymer during its synthesis.Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid has been reported. Electrosensitivity and lower oxidation potential of PPy make it potentially useful for drug delivery, chemical sensors, batteries, ion selective electrodes, biosensor and biochemistry research.5,

Application

PPy may be used to prepare cobalt based electrocatalyst for anion exchange membrane fuel cells.
Conducting polymer.
Conductive additive for thermoplasitics and thermosets.

Packaging

Packaged in glass bottles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XAFS analysis of unpyrolyzed CoPPyC oxygen reduction catalysts for anion-exchange membrane fuel cells (AMFC).
Asazawa K, et al.
Electrochemical Society Transactions, 33(1), 1751-1755 (2010)
Anion-exchange membrane fuel cells: Dual-site mechanism of oxygen reduction reaction in alkaline media on cobalt- polypyrrole electrocatalysts.
Olson TS, et al.
The Journal of Physical Chemistry C, 114(11), 5049-5059 (2010)
In Situ XAFS and HAXPES Analysis and Theoretical Study of Cobalt Polypyrrole Incorporated on Carbon (CoPPyC) Oxygen Reduction Reaction Catalysts for Anion-Exchange Membrane Fuel Cells
Asazawa K, et al.
The Journal of Physical Chemistry C, 118(44), 25480-25486 (2014)
Gusphyl A Justin et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 1206-1209 (2013-02-01)
An electronically-controlled drug delivery system (eDDS) for the on-demand release of anti-inflammatory, anti-microbial and analgesic agents to aid in wound healing is currently under development. The loading of several drugs into conductive polymer films and their subsequent on-demand, controlled release
Lijia Chen et al.
ACS applied materials & interfaces, 5(6), 2047-2052 (2013-03-02)
Herein graphene quantum dot (GQD), a graphene material with lateral dimension less than 100 nm, is explored to dope PPy on F-doped tin oxide glass as an efficient counter electrode for high-performance dye-sensitized solar cells (DSSCs). The GQDs-doped PPy film

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service