Skip to Content
Merck
All Photos(1)

Key Documents

575771

Sigma-Aldrich

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

12.0 wt. %±0.5 wt. % (80% NMP/20% xylene)

Synonym(s):

Pyre-M.L.® RK-692, Polyamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$421.00
1 L
$1,270.00

$421.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$421.00
1 L
$1,270.00

About This Item

Linear Formula:
(C12H12N2O .C10H2O6)x
UNSPSC Code:
12162002
NACRES:
NA.23

$421.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

concentration

12.0 wt. %±0.5 wt. % (80% NMP/20% xylene)

refractive index

n20/D 1.5052

solubility

H2O: miscible

density

1.03 g/mL at 25 °C (relative)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution (PAA) converts to polyimide at 300oC. The FTIR spectra indicates a carbonyl stretch of carboxylic acids in PAA at 1650 cm-1. Following the imidization process, the peak at 1720cm-1 grows, indicating the formation of imide group.[1][2]

Application

It is used as a precursor for polyimide coatings.[3]
It may also be used as a precursor for the preparation of carbon hollow fiber membranes.[4]

Legal Information

Pyre-M.L. is a registered trademark of Industrial Summit Technologies Corp.

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bendable integrated circuits on plastic substrates by use of printed ribbons of single crystalline silicon
Ahn JH
Applied Physics Letters, 90 (2007)
Ultrathin polymer membranes for high throughput CO2 capture
Spadaccini CM
Energy Procedia, 4, 731-736 (2011)
CO2/CH4 mixed gas separation using carbon hollow fiber membranes
Yoshimune M and Haraya K
Energy Procedia, 37, 1109-1116 (2013)
Junping Ju et al.
Journal of colloid and interface science, 404, 36-41 (2013-06-01)
The triblock copolymers with the majority phase comprising fluorinated polyimide and the minor phase consisting of poly (ε-caprolactone) (PCL) were synthesized through Diels-Alder reaction between PI-Maleimide and PCL-Furfuryl Amine. The chemical composition and structure of the copolymers were characterized by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service