All Photos(1)
About This Item
Linear Formula:
(CH3O)2C6H3CH2CH2CO2H
CAS Number:
Molecular Weight:
210.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
solid
mp
66-69 °C (lit.)
functional group
carboxylic acid
SMILES string
COc1ccc(OC)c(CCC(O)=O)c1
InChI
1S/C11H14O4/c1-14-9-4-5-10(15-2)8(7-9)3-6-11(12)13/h4-5,7H,3,6H2,1-2H3,(H,12,13)
InChI key
JENQUCZZZGYHRW-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service